siRNA Details
Gene Symbol: MUC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005960.1 | NP_005951.1 | 12 | 10243 | ||
XM_011516246.2 | XP_011514548.1 | 13 | 10572 | |||
XM_011516247.2 | XP_011514549.1 | 13 | 10522 | |||
XM_011516249.2 | XP_011514551.1 | 13 | 10373 | |||
XM_011516250.2 | XP_011514552.1 | 13 | 9861 | |||
XM_017012231.1 | XP_016867720.1 | 13 | 10069 | |||
GenBank mRNA | AF007194.1 | - | 12 | 3600 | ||
AF143371.1 | - | 10 | 1167 | |||
AF143372.1 | - | 9 | 993 | |||
AF143373.1 | - | 9 | 1051 |
Gene Symbol: MUC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB038783.1 | - | 11 | 2703 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
