siRNA Details
Gene Symbol: U2SURP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080415.1 | NP_001073884.1 | 3, 4 | 314 | ||
NM_001320219.1 | NP_001307148.1 | 3, 4 | 314 | |||
NM_001320220.1 | NP_001307149.1 | 3, 4 | 314 | |||
NM_001320222.1 | NP_001307151.1 | 3, 4 | 314 | |||
XM_005247248.4 | XP_005247305.1 | 3, 4 | 320 | |||
XM_017006038.1 | XP_016861527.1 | 3, 4 | 253 | |||
GenBank mRNA | AB002330.1 | - | 3, 4 | 213 | ||
AK296440.1 | - | 3, 4 | 234 | |||
BC006474.1 | - | 3, 4 | 213 | |||
BC016323.1 | - | 3, 4 | 231 | |||
BC062727.1 | - | 4, 5 | 331 | |||
BC105604.1 | - | 3, 4 | 246 | |||
BC111692.1 | - | 3, 4 | 685 | 40028600 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
