siRNA Details
Gene Symbol: U2SURP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080415.1 | NP_001073884.1 | 4 | 379 | ||
NM_001320219.1 | NP_001307148.1 | 4 | 379 | |||
NM_001320220.1 | NP_001307149.1 | 4 | 379 | |||
NM_001320222.1 | NP_001307151.1 | 4 | 379 | |||
XM_005247248.4 | XP_005247305.1 | 4 | 385 | |||
XM_017006037.1 | XP_016861526.1 | 1 | 58 | |||
XM_017006038.1 | XP_016861527.1 | 4 | 318 | |||
GenBank mRNA | AB002330.1 | - | 4 | 278 | ||
AK296440.1 | - | 4 | 299 | |||
BC006474.1 | - | 4 | 278 | |||
BC016323.1 | - | 4 | 296 | |||
BC062727.1 | - | 5 | 396 | |||
BC105604.1 | - | 4 | 311 | |||
BC111692.1 | - | 4 | 750 | 40028600 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
