siRNA Details
Gene Symbol: LOC100130357 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242698.1 | NP_001229627.1 | 3 | 283 | ||
GenBank mRNA | AK091836.1 | - | 3 | 282 | ||
BX641137.1 | - | 2 | 1666 |
Gene Symbol: TBC1D7-LOC100130357 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134872.1 | 6 | 723 | |||
GenBank mRNA | AK296391.1 | - | 6 | 684 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
