siRNA Details
Gene Symbol: FAM209A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012971.3 | NP_001012989.2 | 1 | 192 | ||
GenBank mRNA | BC029411.1 | - | 1 | 177 | ||
BC070179.1 | - | 1 | 154 | |||
BC107704.1 | - | 1 | 167 | 6614835 | ||
BX335293.2 | - | 1 | 192 |
Gene Symbol: FAM209B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC105792.1 | - | 1, 2 | 173 | 6618487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
