siRNA Details
Gene Symbol: SWI5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040011.1 | NP_001035100.1 | 4 | 587 | ||
NM_001318089.1 | NP_001305018.1 | 4 | 467 | |||
NM_001318092.1 | NP_001305021.1 | 4 | 328 | |||
XM_011518652.2 | XP_011516954.1 | 4 | 602 | |||
GenBank mRNA | AK311408.1 | - | 4 | 492 | ||
BC029911.1 | - | 4 | 511 | |||
BG541668.1 | - | 4 | 370 | |||
CF129530.1 | - | 4 | 328 | |||
DQ414820.1 | - | 4 | 587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
