siRNA Details
Gene Symbol: MBOAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100916.1 | NP_001094386.1 | 3 | 1159 | ||
XM_017013726.1 | XP_016869215.1 | 4 | 1762 | |||
GenBank mRNA | AF359269.1 | - | 1 | 779 | ||
EU518495.2 | - | 3 | 1169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
