siRNA Details
Gene Symbol: LRRC37A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303255.1 | NP_001290184.1 | 11 | 2488 | ||
NM_199340.3 | NP_955372.2 | 15 | 5745 | |||
XM_011524776.1 | XP_011523078.1 | 12 | 3184 | |||
XM_011524777.1 | XP_011523079.1 | 10 | 3031 | |||
XM_011524778.2 | XP_011523080.2 | 13 | 2709 | |||
XM_017024595.1 | XP_016880084.1 | 12 | 5502 | |||
XM_017024597.1 | XP_016880086.1 | 11 | 5421 | |||
XM_017024601.1 | XP_016880090.1 | 12 | 2886 | |||
XM_017024606.1 | XP_016880095.1 | 9 | 2959 | |||
GenBank mRNA | AB011135.2 | - | 14 | 5610 | ||
AK091625.1 | - | 5 | 2593 | |||
AK294371.1 | - | 11 | 2476 | |||
AK299714.1 | - | 8 | 2242 | |||
AK315945.1 | - | 12 | 3217 | |||
BC043145.1 | - | 12 | 5087 | 5296147 |
Gene Symbol: ACBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006717528.3 | XP_006717591.3 | 1 | 2767 | ||
XM_017016884.1 | XP_016872373.1 | 1 | 2767 | |||
XM_017016885.1 | XP_016872374.1 | 1 | 2767 | |||
XM_017016886.1 | XP_016872375.1 | 1 | 2767 | |||
XM_017016890.1 | XP_016872379.1 | 1 | 2767 | |||
XM_017016893.1 | XP_016872382.1 | 1 | 2767 | |||
XM_017016894.1 | XP_016872383.1 | 1 | 2767 |
Gene Symbol: LRRC37A4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002940.2 | 6 | 5189 |
Gene Symbol: LRRC37A6P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003525.2 | 1 | 6399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
