siRNA Details
Gene Symbol: C9orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173520.2 | NP_775791.1 | 2 | 540 | ||
GenBank mRNA | BC034752.1 | - | 2 | 540 | 5193354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
