siRNA Details
Gene Symbol: SMCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015295.2 | NP_056110.2 | 3 | 529 | ||
XM_011525642.1 | XP_011523944.1 | 3 | 529 | |||
XM_011525643.2 | XP_011523945.1 | 3 | 529 | |||
XM_011525644.2 | XP_011523946.1 | 2 | 437 | |||
XM_017025684.1 | XP_016881173.1 | 3 | 529 | |||
GenBank mRNA | BC006008.1 | - | 3 | 529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
