siRNA Details
Gene Symbol: SLC25A53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012755.4 | NP_001012773.2 | 2 | 2625 | ||
XM_005262129.4 | XP_005262186.1 | 2 | 2571 | |||
XM_011530952.2 | XP_011529254.1 | 3 | 2768 | |||
XM_011530953.2 | XP_011529255.1 | 3 | 2599 | |||
XM_017029514.1 | XP_016885003.1 | 3 | 3410 | |||
XM_017029515.1 | XP_016885004.1 | 4 | 2918 | |||
XM_017029516.1 | XP_016885005.1 | 3 | 3541 | |||
GenBank mRNA | AL833609.1 | - | 2 | 2626 | ||
BC140812.1 | - | 1 | 2484 | 9021680 | ||
BC140814.1 | - | 1 | 2485 | 9021682 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
