siRNA Details
Gene Symbol: GCOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY334562.1 | - | 12 | 2020 | ||
AY341345.1 | - | 13 | 2263 | |||
AY353057.1 | - | 14 | 2589 | |||
AY353060.1 | - | 13 | 2496 |
Gene Symbol: POLR2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018102.2 | NP_001018112.1 | 1 | 423 | ||
NM_015532.4 | NP_056347.1 | 1 | 423 | |||
GenBank mRNA | AF326773.1 | - | 1 | 113 | ||
AK074767.1 | - | 1 | 232 | |||
AK074955.1 | - | 1 | 227 | |||
AK128618.1 | - | 1 | 219 | |||
AK223571.1 | - | 1 | 222 | |||
AK308873.1 | - | 1 | 217 | |||
AY339380.1 | - | 1 | 241 | |||
AY341344.1 | - | 1 | 241 | |||
AY353061.1 | - | 1 | 152 | |||
BC001510.2 | - | 1 | 213 | 3504557 | ||
BI833260.1 | - | 1 | 214 | |||
BQ072611.1 | - | 1 | 193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
