siRNA Details
Gene Symbol: RNF151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174903.4 | NP_777563.2 | 1 | 31 | ||
XM_005255129.4 | XP_005255186.1 | 2 | 293 | |||
GenBank mRNA | BC029501.2 | - | 1 | 20 | ||
BC113014.2 | - | 1 | 31 | 40029536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
