siRNA Details
Gene Symbol: FER1L6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040044.1 | 4 | 552 | |||
GenBank mRNA | AK093040.1 | - | 4 | 552 | ||
BC137357.1 | - | 4 | 519 | 9021355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
