siRNA Details
Gene Symbol: MLLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005934.3 | NP_005925.2 | 12 | 2852 | ||
XM_011528021.1 | XP_011526323.1 | 12 | 2826 | |||
XM_011528022.1 | XP_011526324.1 | 12 | 2709 | |||
XM_011528023.1 | XP_011526325.1 | 11 | 2700 | |||
XM_017026819.1 | XP_016882308.1 | 12 | 3408 | |||
GenBank mRNA | AL365410.1 | - | 1 | 197 | ||
BC047008.1 | - | 1 | 387 | |||
BC084564.1 | - | 1 | 358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
