siRNA Details
Gene Symbol: GPR137C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099652.1 | NP_001093122.1 | 7 | 2390 | ||
GenBank mRNA | AL834372.1 | - | 8 | 2296 | ||
BX647179.1 | - | 5 | 3911 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
