siRNA Details
Gene Symbol: GPR137C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099652.1 | NP_001093122.1 | 3 | 706 | ||
GenBank mRNA | AK123953.1 | - | 3 | 602 | ||
AK293812.1 | - | 1 | 155 | |||
AL834372.1 | - | 3 | 566 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
