siRNA Details
Gene Symbol: FAM109B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002034.2 | NP_001002034.2 | 3 | 1379 | ||
XM_005261373.3 | XP_005261430.1 | 3 | 1517 | |||
GenBank mRNA | BC029776.1 | - | 1 | 1033 | ||
BX648402.1 | - | 3 | 1379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
