siRNA Details
Gene Symbol: C8orf59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099670.2 | NP_001093140.1 | 3 | 350 | ||
NM_001099671.2 | NP_001093141.1 | 3 | 347 | |||
NM_001099672.2 | NP_001093142.1 | 3 | 263 | |||
NM_001099673.2 | NP_001093143.1 | 2 | 222 | |||
NM_001293320.1 | NP_001280249.1 | 2 | 222 | |||
NR_120681.1 | 3 | 263 | ||||
XM_011517528.2 | XP_011515830.1 | 3 | 247 | |||
GenBank mRNA | AK000414.1 | - | 171 | |||
BC032347.1 | - | 3 | 180 | |||
BC055092.1 | - | 2 | 178 | |||
BC062221.1 | - | 3 | 219 | |||
BC146983.1 | - | 2 | 167 | 9056911 | ||
BC146988.1 | - | 2 | 167 | 9056916 | ||
BC157825.1 | - | 2 | 167 | 9057044 | ||
BC171873.1 | - | 2 | 166 | |||
BG926519.1 | - | 363 | ||||
BM785759.1 | - | 3 | 302 | |||
CB119146.1 | - | 2 | 156 | |||
CN280666.1 | - | 3 | 190 | |||
CN481353.1 | - | 2 | 221 | |||
DB094478.1 | - | 2 | 214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
