siRNA Details
Gene Symbol: LINC01312 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027030.1 | 1, 2 | 599 | |||
GenBank mRNA | BC032958.2 | - | 1, 2 | 599 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
