siRNA Details
Gene Symbol: GUSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027026.1 | 4, 5 | 390 | |||
GenBank mRNA | AK095656.1 | - | 4, 5 | 314 | ||
DA358891.1 | - | 4, 5 | 390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
