siRNA Details
Gene Symbol: IGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123975.1 | - | 5 | 1132 | ||
AK125079.1 | - | 4 | 1095 | |||
AK126352.1 | - | 5 | 1078 | |||
BC066594.1 | - | 5 | 1041 | |||
BX538066.1 | - | 6 | 1027 | |||
BX640822.1 | - | 5 | 1121 | |||
BX640854.1 | - | 5 | 1109 |
Gene Symbol: IGHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL389978.1 | - | 4 | 842 | ||
BC073765.1 | - | 4 | 1049 | |||
BX538077.1 | - | 5 | 1070 | |||
BX640625.1 | - | 5 | 1092 | |||
BX640710.1 | - | 4 | 1087 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
