siRNA Details
Gene Symbol: IGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123975.1 | - | 5 | 1135 | ||
AK125079.1 | - | 4 | 1098 | |||
AK126352.1 | - | 5 | 1081 | |||
BC066594.1 | - | 5 | 1044 | |||
BX538066.1 | - | 6 | 1030 | |||
BX640822.1 | - | 5 | 1124 | |||
BX640854.1 | - | 5 | 1112 |
Gene Symbol: IGHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL389978.1 | - | 4 | 845 | ||
BC073765.1 | - | 4 | 1052 | |||
BX538077.1 | - | 5 | 1073 | |||
BX640625.1 | - | 5 | 1095 | |||
BX640710.1 | - | 4 | 1090 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
