siRNA Details
Gene Symbol: CLIC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114086.1 | NP_001107558.1 | 6 | 4773 | ||
NM_016929.4 | NP_058625.2 | 6 | 4537 | |||
NR_045672.1 | 3 | 4115 | ||||
NR_045673.1 | 5 | 4189 | ||||
XM_017010953.1 | XP_016866442.1 | 7 | 5018 | |||
GenBank mRNA | AJ420547.1 | - | 1 | 151 | ||
AK025350.1 | - | 1 | 878 | |||
AL049313.1 | - | 1 | 993 | |||
BC020923.1 | - | 1 | 301 | |||
DQ679794.1 | - | 6 | 4505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
