siRNA Details
Gene Symbol: CLIC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114086.1 | NP_001107558.1 | 6 | 4640 | ||
NM_016929.4 | NP_058625.2 | 6 | 4404 | |||
NR_045672.1 | 3 | 3982 | ||||
NR_045673.1 | 5 | 4056 | ||||
XM_017010953.1 | XP_016866442.1 | 7 | 4885 | |||
GenBank mRNA | AK025350.1 | - | 1 | 745 | ||
AL049313.1 | - | 1 | 860 | |||
BC020923.1 | - | 1 | 168 | |||
DQ679794.1 | - | 6 | 4372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
