siRNA Details
Gene Symbol: KIF5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB210045.1 | - | 29 | 5543 | ||
AB290174.1 | - | 29 | 5784 | |||
BC033961.2 | - | 1 | 455 | |||
BC051752.1 | - | 1 | 1760 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
