siRNA Details
Gene Symbol: ZNF836 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102657.1 | NP_001096127.1 | 5 | 1723 | ||
XM_011526558.2 | XP_011524860.1 | 5 | 1565 | |||
XM_011526559.2 | XP_011524861.1 | 4 | 1610 | |||
XM_017026389.1 | XP_016881878.1 | 5 | 1498 | |||
GenBank mRNA | AK131308.1 | - | 5 | 1575 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
