siRNA Details
Gene Symbol: MYPOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012643.3 | NP_001012661.1 | 3 | 1787 | ||
XM_017026742.1 | XP_016882231.1 | 2 | 2532 | |||
XM_017026743.1 | XP_016882232.1 | 3 | 2642 | |||
GenBank mRNA | BC044311.1 | - | 3 | 1787 | 5756168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
