siRNA Details
Gene Symbol: CSNK1A1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027320.1 | 4 | 1077 | |||
GenBank mRNA | BC028192.1 | - | 4 | 1030 | ||
BC066972.1 | - | 4 | 868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
