siRNA Details
Gene Symbol: BREA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015445.1 | 2 | 966 | |||
GenBank mRNA | AL833948.1 | - | 2 | 966 | ||
AY940073.1 | - | 2 | 1935 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
