siRNA Details
Gene Symbol: ZFP90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305203.1 | NP_001292132.1 | 5 | 2673 | ||
NM_001305206.1 | NP_001292135.1 | 5 | 2728 | |||
NM_001305207.1 | NP_001292136.1 | 5 | 2725 | |||
NM_001305208.1 | NP_001292137.1 | 5 | 2718 | |||
NM_133458.3 | NP_597715.2 | 5 | 2663 | |||
NR_130976.1 | 5 | 2771 | ||||
NR_130977.1 | 5 | 2721 | ||||
NR_130978.1 | 5 | 2779 | ||||
XM_005255804.2 | XP_005255861.1 | 5 | 2603 | |||
XM_017022952.1 | XP_016878441.1 | 4 | 2710 | |||
GenBank mRNA | AB075834.1 | - | 1 | 1870 | ||
AK056596.1 | - | 1 | 1459 | |||
AK074332.1 | - | 5 | 2687 | |||
AK122847.1 | - | 1 | 3101 | |||
AK123121.1 | - | 1 | 319 | |||
AK126167.1 | - | 2 | 875 | |||
BC033165.1 | - | 1 | 1359 | |||
BC040486.1 | - | 5 | 2676 | |||
BC140886.1 | - | 5 | 2666 | 9021754 | ||
BC140887.1 | - | 5 | 2666 | 9021755 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
