siRNA Details
Gene Symbol: ZFP90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305203.1 | NP_001292132.1 | 5 | 4171 | ||
NM_001305206.1 | NP_001292135.1 | 5 | 4226 | |||
NM_001305207.1 | NP_001292136.1 | 5 | 4223 | |||
NM_001305208.1 | NP_001292137.1 | 5 | 4216 | |||
NM_133458.3 | NP_597715.2 | 5 | 4161 | |||
NR_130976.1 | 5 | 4269 | ||||
NR_130977.1 | 5 | 4219 | ||||
NR_130978.1 | 5 | 4277 | ||||
XM_005255804.2 | XP_005255861.1 | 5 | 4101 | |||
XM_017022952.1 | XP_016878441.1 | 4 | 4208 | |||
GenBank mRNA | AB075834.1 | - | 1 | 3373 | ||
AK123121.1 | - | 1 | 1821 | |||
AK126167.1 | - | 2 | 2376 | |||
BC040486.1 | - | 5 | 4177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
