siRNA Details
Gene Symbol: GCC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135290.1 | 2 | 444 | |||
GenBank mRNA | AK095987.1 | - | 2 | 444 | ||
BC004487.2 | - | 2 | 421 | 3832541 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
