siRNA Details
Gene Symbol: TOB2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002936.2 | 1 | 478 | |||
GenBank mRNA | AK292767.1 | - | 1 | 435 | ||
BC015543.1 | - | 1 | 498 | |||
BC039027.1 | - | 1 | 516 | |||
BC050562.1 | - | 1 | 478 | |||
BC126996.1 | - | 1 | 40 | |||
BC126997.1 | - | 1 | 40 | |||
HQ268503.1 | - | 1 | 40 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
