siRNA Details
Gene Symbol: UFSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015072.3 | NP_001015072.2 | 1 | 707 | ||
GenBank mRNA | AK292467.1 | - | 1 | 707 | ||
BC017456.2 | - | 1 | 642 | |||
BC067904.1 | - | 1 | 670 | 5310615 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
