siRNA Details
Gene Symbol: ST20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100879.1 | NP_001094349.1 | 3 | 749 | ||
NM_001100880.2 | NP_001094350.1 | 3 | 282 | |||
NM_001199757.1 | NP_001186686.1 | 3 | 260 | |||
NR_037652.1 | 3 | 383 | ||||
NR_037653.1 | 3 | 405 | ||||
GenBank mRNA | AA282293.1 | - | 3 | 307 | ||
AF249277.1 | - | 3 | 351 | |||
AL109714.1 | - | 3 | 749 | |||
BM559302.1 | - | 3 | 372 | |||
BQ680929.1 | - | 3 | 364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
