siRNA Details
Gene Symbol: CEACAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039213.3 | NP_001034302.2 | 5 | 938 | ||
XM_017026795.1 | XP_016882284.1 | 4 | 1633 | |||
GenBank mRNA | BC144608.1 | - | 5 | 938 | ||
EU021223.1 | - | 4 | 732 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
