siRNA Details
Gene Symbol: CCDC168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146197.1 | NP_001139669.1 | 4 | 20102 | ||
XM_011521106.1 | XP_011519408.1 | 5 | 19975 | |||
GenBank mRNA | AK125984.1 | - | 1 | 1965 | ||
AL833499.1 | - | 1 | 3850 | |||
AL833908.1 | - | 1 | 45 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
