siRNA Details
Gene Symbol: PLEKHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161498.1 | NP_001154970.1 | 8 | 893 | ||
XM_011536762.2 | XP_011535064.1 | 5 | 339 | |||
XM_011536763.2 | XP_011535065.1 | 3 | 199 | |||
XM_017021290.1 | XP_016876779.1 | 8 | 1255 | |||
GenBank mRNA | BC146872.1 | - | 8 | 798 | 9056800 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
