siRNA Details
Gene Symbol: ESR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122741.1 | NP_001116213.1 | 1 | 114 | ||
NM_001291230.1 | NP_001278159.1 | 1 | 114 | |||
NM_001291241.1 | NP_001278170.1 | 1 | 114 | |||
GenBank mRNA | AK293019.1 | - | 1 | 82 | ||
AK313644.1 | - | 1 | 82 | |||
DB285906.1 | - | 1 | 114 | |||
X74439.1 | - | 1 | 1375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
