siRNA Details
Gene Symbol: CEP295NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243540.1 | NP_001230469.1 | 3 | 360 | ||
NM_001243541.1 | NP_001230470.1 | 3 | 421 | |||
GenBank mRNA | AK057217.1 | - | 3 | 344 | ||
DB037087.1 | - | 3 | 421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
