siRNA Details
Gene Symbol: PLPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005269592.2 | XP_005269649.1 | 6 | 10368 | ||
XM_006717685.3 | XP_006717748.1 | 7 | 10208 | |||
XM_011539444.2 | XP_011537746.1 | 8 | 10266 | |||
XM_017015821.1 | XP_016871310.1 | 4 | 9949 | |||
XM_017015822.1 | XP_016871311.1 | 4 | 9916 |
Gene Symbol: LINC01561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103717.1 | 1 | 925 | |||
GenBank mRNA | AK094721.1 | - | 1 | 665 | ||
BC108662.1 | - | 1 | 925 | 3922405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
