siRNA Details
Gene Symbol: LINC00602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027284.1 | 1 | 1267 | |||
GenBank mRNA | AK094116.1 | - | 1 | 1267 | ||
BC110806.1 | - | 1 | 1191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
