siRNA Details
Gene Symbol: CLDN22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111319.1 | NP_001104789.1 | 1 | 1194 | ||
GenBank mRNA | AK098064.1 | - | 1 | 588 | ||
AK303864.1 | - | 648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
