siRNA Details
Gene Symbol: MTUS2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046378.1 | 2 | 337 | |||
GenBank mRNA | BC027486.1 | - | 2 | 337 | ||
BI223246.1 | - | 2 | 336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
