siRNA Details
Gene Symbol: ANKRD34A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039888.3 | NP_001034977.1 | 4 | 2874 | ||
GenBank mRNA | AK094282.1 | - | 2 | 1775 | ||
AK128050.1 | - | 4 | 2868 | |||
AL359622.1 | - | 1 | 554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
