siRNA Details
Gene Symbol: GNA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002067.4 | NP_002058.2 | 7 | 3695 | ||
GenBank mRNA | AL110227.1 | - | 1 | 2043 | ||
BC001528.2 | - | 1, 2 | 1095 | |||
BC051315.1 | - | 1 | 950 | |||
BC111027.1 | - | 1 | 1133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
