siRNA Details
Gene Symbol: WDR97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316309.1 | NP_001303238.1 | 9, 10 | 2357 | ||
XM_017013385.1 | XP_016868874.1 | 9, 10 | 2351 | |||
XM_017013386.1 | XP_016868875.1 | 9, 10 | 2351 | |||
XM_017013387.1 | XP_016868876.1 | 9, 10 | 2292 | |||
XM_017013388.1 | XP_016868877.1 | 9, 10 | 2351 | |||
XM_017013389.1 | XP_016868878.1 | 9, 10 | 2351 | |||
XM_017013390.1 | XP_016868879.1 | 9, 10 | 2351 | |||
XM_017013391.1 | XP_016868880.1 | 9, 10 | 2351 | |||
XM_017013392.1 | XP_016868881.1 | 9, 10 | 2351 | |||
XM_017013393.1 | XP_016868882.1 | 9, 10 | 2351 | |||
XM_017013394.1 | XP_016868883.1 | 8, 9 | 2137 | |||
GenBank mRNA | AB058778.1 | - | 9, 10 | 2323 | ||
AK095597.1 | - | 3, 4 | 1089 | |||
AK302485.1 | - | 5, 6 | 878 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
