siRNA Details
Gene Symbol: HSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080439.2 | NP_001073908.2 | 4 | 1512 | ||
XM_011524283.1 | XP_011522585.1 | 4 | 1525 | |||
GenBank mRNA | BC033017.1 | - | 4 | 1476 | 4825807 | |
BC033020.1 | - | 4 | 1499 | 4823233 | ||
BC115385.1 | - | 5 | 1088 | 40038134 | ||
BC115386.1 | - | 5 | 1084 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
