siRNA Details
Gene Symbol: GK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000167.5 | NP_000158.1 | 2, 3 | 338 | ||
NM_001128127.2 | NP_001121599.1 | 2, 3 | 338 | |||
NM_001205019.1 | NP_001191948.1 | 2, 3 | 338 | |||
NM_203391.3 | NP_976325.1 | 2, 3 | 338 | |||
XM_006724483.2 | XP_006724546.1 | 2, 3 | 291 | |||
XM_006724484.2 | XP_006724547.1 | 2, 3 | 290 | |||
XM_011545491.2 | XP_011543793.1 | 2, 3 | 292 | |||
XM_011545492.1 | XP_011543794.1 | 2, 3 | 291 | |||
XM_017029409.1 | XP_016884898.1 | 2, 3 | 208 | |||
GenBank mRNA | AK294938.1 | - | 2, 3 | 208 | ||
AK313215.1 | - | 2, 3 | 274 | |||
BC037549.1 | - | 2, 3 | 195 | 5213716 | ||
BC042421.1 | - | 2, 3 | 339 | 4820945 | ||
BC071595.1 | - | 2, 3 | 339 | 5276056 | ||
BM906503.1 | - | 285 | ||||
BX647604.1 | - | 2, 3 | 199 | |||
L13943.1 | - | 2, 3 | 225 |
Gene Symbol: GK3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026575.1 | 1 | 537 | |||
GenBank mRNA | AK225740.1 | - | 1 | 188 | ||
AK292282.1 | - | 1 | 537 | |||
BC066960.1 | - | 1 | 193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
